Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INSIG1
Entrez Gene ID
403162
Gene Name
insulin induced gene 1
Gene Aliases
CL-6
Chromosome Location
Chr.18: 2885494 - 2898758 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244521.1 NP_001231450.1 5 - 6 897 64
GenBank mRNA AK237645.1 - 5 - 6 906 64
FD641154.1 - 4 - 5 451 64

View Full Product Details