Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FKBP14
Entrez Gene ID
100514551
Gene Name
FKBP prolyl isomerase 14
Gene Aliases
-
Chromosome Location
Chr.18: 42903100 - 42913609 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244527.1 NP_001231456.1 2 - 3 517 103
GenBank mRNA AY610280.1 - 2 - 3 351 103
EW454624.2 - 2 - 3 358 103
FD642407.1 - 2 - 3 517 103

View Full Product Details