Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRT27
Entrez Gene ID
100521040
Gene Name
keratin 27
Gene Aliases
-
Chromosome Location
Chr.12: 21677109 - 21682465 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244622.1 NP_001231551.1 3 - 4 735 90
GenBank mRNA AK349825.1 - 3 - 4 735 90
AK397166.1 - 3 - 4 738 90
AK397400.1 - 3 - 4 732 90

View Full Product Details