Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MADS2
Entrez Gene ID
100232867
Gene Name
MADS-box protein 2
Gene Aliases
-
Chromosome Location
Chr.14: 12561660 - 12577986 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF373601.1 - 8 - 9 751 76

View Full Product Details