Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MADS3
Entrez Gene ID
100232868
Gene Name
MADS-box protein 3
Gene Aliases
GSVIVT00026310001
Chromosome Location
Chr.15: 5979042 - 5994124 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF373602.1 - 3 - 4 417 88

View Full Product Details