Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIP2;1
Entrez Gene ID
100233002
Gene Name
aquaporin-like
Gene Aliases
PIP2-1, aquaporin
Chromosome Location
Chr.13: 5706931 - 5709930 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY823263.1 - 1 - 2 373 71
EF364438.1 - 1 - 2 312 71

View Full Product Details