Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAR2
Entrez Gene ID
100233112
Gene Name
leucoanthocyanidin reductase 2
Gene Aliases
GSVIVT00017628001
Chromosome Location
Chr.17: 4270654 - 4273659 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA DQ129686.1 - 3 - 4 611 92

View Full Product Details