Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WRKY2
Entrez Gene ID
100233123
Gene Name
DNA binding protein WRKY2
Gene Aliases
-
Chromosome Location
Chr.Un_NW_002240771: 39790 - 53755 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY596466.2 - 1 - 2 477 58

View Full Product Details