Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIP1;1
Entrez Gene ID
100233003
Gene Name
aquaporin-like
Gene Aliases
GSVIVT00018548001, aquaporin
Chromosome Location
Chr.13: 2514309 - 2516070 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY839872.2 - 2 - 3 430 95
DQ834701.1 - 2 - 3 449 95

View Full Product Details