Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RD22-B
Entrez Gene ID
100242354
Gene Name
rd22-b protein
Gene Aliases
-
Chromosome Location
Chr.11: 3197012 - 3198437 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FJ869893.2 - 1 - 2 41 62

View Full Product Details