Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CBL07
Entrez Gene ID
100254259
Gene Name
calcineurin B-like protein 07
Gene Aliases
GSVIVT00032178001
Chromosome Location
Chr.4: 843202 - 849012 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FJ901273.1 - 8 - 9 700 101

View Full Product Details