Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CBL08
Entrez Gene ID
100254683
Gene Name
calcineurin B-like protein 08
Gene Aliases
GSVIVT00006734001
Chromosome Location
Chr.2: 1442663 - 1448961 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FJ901274.1 - 2 - 3 174 81

View Full Product Details