Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
myh4
Entrez Gene ID
407946
Gene Name
myosin, heavy chain 4, skeletal muscle
Gene Aliases
E3, mhc, myh2b, myhc-2b, myhc-iib, myhe3
Chromosome Location
Chr.8: 83534466 - 83548537 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001244.1 NP_001001244.1 29 - 30 4213 94
GenBank mRNA BC067305.1 - 29 - 30 4213 94

View Full Product Details