The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001244.1 | NP_001001244.1 | 6 - 7 | 688 | 66 |
GenBank mRNA | BC067305.1 | - | 6 - 7 | 688 | 66 |