Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cdca8
Entrez Gene ID
444885
Gene Name
cell division cycle associated 8
Gene Aliases
Dasra-B, bor, borealin, dasrab
Chromosome Location
Chr.2: 58938238 - 58954643 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002902.1 NP_001002902.1 5 - 6 396 87
XM_012956694.2 XP_012812148.1 6 - 7 87
XM_018091156.1 XP_017946645.1 5 - 6 87
GenBank mRNA BC074572.1 - 5 - 6 396 87
CR762304.2 - 4 - 5 87

View Full Product Details