Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cdca8
Entrez Gene ID
444885
Gene Name
cell division cycle associated 8
Gene Aliases
Dasra-B, bor, borealin, dasrab
Chromosome Location
Chr.2: 58938238 - 58954643 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002902.1 NP_001002902.1 8 - 9 647 92
XM_012956694.2 XP_012812148.1 9 - 10 856 92
XM_018091156.1 XP_017946645.1 8 - 9 694 92
GenBank mRNA BC074572.1 - 8 - 9 647 92
CR762304.2 - 7 - 8 598 92

View Full Product Details