Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rac1
Entrez Gene ID
448118
Gene Name
Rac family small GTPase 1
Gene Aliases
Xrac, mig5, p21-rac1, rac, tc-25
Chromosome Location
Chr.9: 19616457 - 19631401 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004840.1 NP_001004840.1 5 - 6 620 57
GenBank mRNA BC074649.1 - 5 - 6 620 57
CR760977.2 - 5 - 6 569 57

View Full Product Details