Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
btn2a2
Entrez Gene ID
448245
Gene Name
butyrophilin subfamily 2 member A2
Gene Aliases
bt2.2, btf2
Chromosome Location
Chr.Un|NW_016683613: 45176 - 63184 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004895.1 NP_001004895.1 - 946 90
GenBank mRNA BC075308.1 - - 946 90

View Full Product Details