Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sult1c2
Entrez Gene ID
448251
Gene Name
sulfotransferase family 1C member 2
Gene Aliases
st1c1, st1c2, sult1c1
Chromosome Location
Chr.Un|NW_016683961: 182201 - 188084 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004900.1 NP_001004900.1 4 - 5 817 98
XM_012954910.2 XP_012810364.1 4 - 5 2369 98
GenBank mRNA BC075315.1 - 4 - 5 817 98

View Full Product Details