Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pdzd11
Entrez Gene ID
448266
Gene Name
PDZ domain containing 11
Gene Aliases
-
Chromosome Location
Chr.8: 17760373 - 17766399 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004906.1 NP_001004906.1 5 - 6 440 96
XM_018096199.1 XP_017951688.1 4 - 5 420 96
GenBank mRNA BC075334.1 - 5 - 6 440 96

View Full Product Details