Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rnd3
Entrez Gene ID
448301
Gene Name
Rho family GTPase 3
Gene Aliases
arhe, memb, rho8, rhoe
Chromosome Location
Chr.9: 71415689 - 71438748 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004920.1 NP_001004920.1 1 - 2 259 87
GenBank mRNA BC075375.1 - 1 - 2 259 87

View Full Product Details