Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dynll1
Entrez Gene ID
448652
Gene Name
dynein light chain LC8-type 1
Gene Aliases
dlc1, dlc8, dlc8a, dncl1, dnclc1, dynll1-a, dynll1-b, hdlc1, lc8, lc8a, pin
Chromosome Location
Chr.1: 143209439 - 143211054 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005077.1 NP_001005077.1 2 - 3 202 102
GenBank mRNA BC076999.1 - 2 - 3 202 102
CR761866.2 - 2 - 3 187 102

View Full Product Details