Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc30a8
Entrez Gene ID
496450
Gene Name
solute carrier family 30 (zinc transporter), member 8
Gene Aliases
slc30a3, znt8
Chromosome Location
Chr.6: 121687257 - 121713950 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011041.1 NP_001011041.1 6 - 7 974 72
XM_018094625.1 XP_017950114.1 5 - 6 983 72
GenBank mRNA BC084148.1 - 6 - 7 974 72

View Full Product Details