Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kdelr2
Entrez Gene ID
496456
Gene Name
KDEL endoplasmic reticulum protein retention receptor 2
Gene Aliases
kdelr1
Chromosome Location
Chr.7: 107818848 - 107842469 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011046.1 NP_001011046.1 2 - 3 298 93
GenBank mRNA BC084170.1 - 2 - 3 298 93
CR761657.2 - 2 - 3 328 93

View Full Product Details