Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rbm18
Entrez Gene ID
496464
Gene Name
RNA binding motif protein 18
Gene Aliases
-
Chromosome Location
Chr.8: 16299005 - 16310106 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011054.1 NP_001011054.1 4 - 5 370 87
GenBank mRNA BC084179.1 - 4 - 5 370 87

View Full Product Details