Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tardbp
Entrez Gene ID
394651
Gene Name
TAR DNA binding protein
Gene Aliases
TDP-43, tdp43
Chromosome Location
Chr.7: 39618757 - 39629202 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244759.1 NP_001231688.1 3 - 4 500 103
NM_203723.2 NP_989054.2 5 - 6 914 103
XM_012966272.2 XP_012821726.1 5 - 6 900 103
GenBank mRNA AL659317.2 - 3 - 4 463 103
BC061336.1 - 5 - 6 869 103
CR762161.2 - 5 - 6 813 103

View Full Product Details