Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
cldn7
Entrez Gene ID
394674
Gene Name
claudin 7
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016684566: 6938 - 12058 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203746.1 NP_989077.1 - 414 103
GenBank mRNA BC061261.1 - - 414 103
CT025424.2 - 1 - 2 328 103

View Full Product Details