Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
arglu1
Entrez Gene ID
394894
Gene Name
arginine and glutamate rich 1
Gene Aliases
-
Chromosome Location
Chr.2: 114925154 - 114937866 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203949.1 NP_989280.1 1 - 2 403 82
GenBank mRNA BC064271.1 - 1 - 2 403 82

View Full Product Details