Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tgif1
Entrez Gene ID
395014
Gene Name
TGFB induced factor homeobox 1
Gene Aliases
hpe4, tgif
Chromosome Location
Chr.6: 91802774 - 91815903 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204051.1 NP_989382.1 2 - 3 542 57
XM_012964471.2 XP_012819925.1 2 - 3 586 57
XM_012964472.2 XP_012819926.1 2 - 3 344 57
GenBank mRNA BC064716.1 - 2 - 3 542 57
CR848339.2 - 2 - 3 547 57

View Full Product Details