Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fkbp14
Entrez Gene ID
448687
Gene Name
FK506 binding protein 14
Gene Aliases
-
Chromosome Location
Chr.6: 80479367 - 80486474 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005108.1 NP_001005108.1 2 - 3 495 107
GenBank mRNA BC077042.1 - 2 - 3 495 107
CR761056.2 - 2 - 3 443 107

View Full Product Details