Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wdr74
Entrez Gene ID
448734
Gene Name
WD repeat domain 74
Gene Aliases
-
Chromosome Location
Chr.4: 41178710 - 41186690 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005144.1 NP_001005144.1 2 - 3 174 99
GenBank mRNA BC077693.1 - 2 - 3 174 99
CR761697.2 - 2 - 3 179 99

View Full Product Details