Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rlbp1
Entrez Gene ID
448050
Gene Name
retinaldehyde binding protein 1
Gene Aliases
CRALBP
Chromosome Location
Chr.3: 99633573 - 99646918 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005455.1 NP_001005455.1 5 - 6 641 116
GenBank mRNA BC074571.1 - 5 - 6 641 116
CR855832.2 - 5 - 6 587 116

View Full Product Details