Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
agr3
Entrez Gene ID
448756
Gene Name
anterior gradient 3, protein disulphide isomerase family member
Gene Aliases
ag3, agr2, bcmp11, hag-3, hag3, pdia18
Chromosome Location
Chr.6: 44522412 - 44534247 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006908.1 NP_001006909.1 6 - 7 480 99
XM_012964546.2 XP_012820000.1 6 - 7 491 99
GenBank mRNA BC075325.1 - 6 - 7 480 99

View Full Product Details