Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tbl2
Entrez Gene ID
493446
Gene Name
transducin (beta)-like 2
Gene Aliases
wbscr13, ws-betatrp
Chromosome Location
Chr.2: 31770230 - 31778149 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008083.1 NP_001008084.1 3 - 4 522 87
GenBank mRNA BC080983.1 - 3 - 4 522 87
CR926407.2 - 3 - 4 440 87

View Full Product Details