Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ttpa
Entrez Gene ID
493547
Gene Name
tocopherol (alpha) transfer protein
Gene Aliases
-
Chromosome Location
Chr.6: 102973668 - 102984135 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008184.1 NP_001008185.1 4 - 5 697 68
GenBank mRNA BC082509.1 - 4 - 5 697 68

View Full Product Details