Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lasp1
Entrez Gene ID
493322
Gene Name
LIM and SH3 protein 1
Gene Aliases
lasp-1, mln50
Chromosome Location
Chr.10: 3679194 - 3713100 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008445.1 NP_001008445.1 1 - 2 288 80
GenBank mRNA BC080453.1 - 1 - 2 288 80
CR855667.2 - 1 - 2 200 80

View Full Product Details