Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ywhab
Entrez Gene ID
496529
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta
Gene Aliases
14-3-3beta, gw128, hs1, kcip-1, ywhab-a, ywhab-b
Chromosome Location
Chr.10: 5520254 - 5531371 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011116.1 NP_001011116.1 3 - 4 518 80
GenBank mRNA BC084514.1 - 3 - 4 518 80
CR760847.2 - 3 - 4 493 80

View Full Product Details