Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
otud6b
Entrez Gene ID
496720
Gene Name
OTU domain containing 6B
Gene Aliases
duba5
Chromosome Location
Chr.6: 111755630 - 111763730 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011269.1 NP_001011269.1 2 - 3 284 75
GenBank mRNA BC087978.1 - 2 - 3 284 75

View Full Product Details