Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cdk1
Entrez Gene ID
394503
Gene Name
cyclin-dependent kinase 1
Gene Aliases
MPF, PSTAIR, cdc-2, cdc2, cdc28a, cdc2x1.1, cdk1-a, cdk1-b, p34cdc2, xcdc2
Chromosome Location
Chr.7: 8473586 - 8481982 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203577.1 NP_988908.1 4 - 5 402 88
XM_012966262.1 XP_012821716.1 4 - 5 438 88
XM_012966263.2 XP_012821717.1 4 - 5 410 88
GenBank mRNA BC061617.1 - 4 - 5 402 88
BC077651.1 - 4 - 5 374 88
CR762071.2 - 3 - 4 338 88

View Full Product Details