Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mtap
Entrez Gene ID
496996
Gene Name
methylthioadenosine phosphorylase
Gene Aliases
-
Chromosome Location
Chr.1: 108956955 - 108970356 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011495.1 NP_001011495.1 2 - 3 123 116
XM_012961488.1 XP_012816942.1 2 - 3 120 116
XM_018092899.1 XP_017948388.1 2 - 3 956 116
GenBank mRNA BC088885.1 - 2 - 3 123 116
CR762293.2 - 2 - 3 106 116

View Full Product Details