Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rps23
Entrez Gene ID
497003
Gene Name
ribosomal protein S23
Gene Aliases
-
Chromosome Location
Chr.1: 168219682 - 168223110 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011499.1 NP_001011499.1 2 - 3 298 81
GenBank mRNA BC088894.1 - 2 - 3 298 81
CR942723.2 - - 288 81

View Full Product Details