Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ctrl
Entrez Gene ID
548358
Gene Name
chymotrypsin-like
Gene Aliases
ctrb1
Chromosome Location
Chr.4: 43576111 - 43582299 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015686.1 NP_001015686.1 5 - 6 501 75
GenBank mRNA BC089075.1 - 5 - 6 501 75

View Full Product Details