Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dcaf10
Entrez Gene ID
548459
Gene Name
DDB1 and CUL4 associated factor 10
Gene Aliases
wdr32
Chromosome Location
Chr.1: 96538529 - 96546148 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015742.2 NP_001015742.2 2 - 3 552 75
GenBank mRNA BC089677.1 - 2 - 3 526 75
BC159065.1 - 2 - 3 540 75
BC159294.1 - 2 - 3 531 75
BC170811.1 - 2 - 3 267 75
BC170813.1 - 2 - 3 267 75
CF149630.1 - 2 - 3 552 75
CR762027.2 - 2 - 3 452 75

View Full Product Details