Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dcaf10
Entrez Gene ID
548459
Gene Name
DDB1 and CUL4 associated factor 10
Gene Aliases
wdr32
Chromosome Location
Chr.1: 96538529 - 96546148 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015742.2 NP_001015742.2 6 - 7 1210 74
GenBank mRNA BC089677.1 - 6 - 7 1184 74
BC159065.1 - 6 - 7 1198 74
BC159294.1 - 6 - 7 1189 74
BC170811.1 - 6 - 7 925 74
BC170813.1 - 6 - 7 925 74
CR762027.2 - 6 - 7 1110 74

View Full Product Details