Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cxcl12
Entrez Gene ID
548481
Gene Name
C-X-C motif chemokine ligand 12
Gene Aliases
pbsf, scyb12, sdf-1, sdf-1a, sdf-1b, sdf1, sdf1a, sdf1b, tlsf-a, tlsf-b, tpar1, xSDF-1, xSDF-1alpha
Chromosome Location
Chr.7: 37782393 - 37841001 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015764.1 NP_001015764.1 3 - 4 288 90
GenBank mRNA BC089703.1 - 3 - 4 288 90

View Full Product Details