Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
c2orf40
Entrez Gene ID
548499
Gene Name
chromosome 2 open reading frame 40
Gene Aliases
ecrg4
Chromosome Location
Chr.2: 106493503 - 106507664 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015782.1 NP_001015782.2 3 - 4 313 64
GenBank mRNA BC089724.1 - 3 - 4 313 64

View Full Product Details