Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aph1a
Entrez Gene ID
548560
Gene Name
aph-1 homolog A, gamma secretase subunit
Gene Aliases
aph-1a, cgi-78
Chromosome Location
Chr.8: 111973822 - 111981772 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015843.1 NP_001015843.1 6 - 7 757 58
GenBank mRNA BC090132.1 - 6 - 7 757 58
CT030063.1 - 6 - 7 751 58

View Full Product Details