Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dusp5
Entrez Gene ID
548573
Gene Name
dual specificity phosphatase 5
Gene Aliases
dusp, hvh3
Chromosome Location
Chr.Un|NW_016683658: 101779 - 109540 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015856.1 NP_001015856.1 2 - 3 514 64
GenBank mRNA BC090366.1 - 2 - 3 514 64

View Full Product Details