Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zdhhc21
Entrez Gene ID
548827
Gene Name
zinc finger DHHC-type containing 21
Gene Aliases
-
Chromosome Location
Chr.1: 111989753 - 112009471 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016073.2 NP_001016073.1 5 - 6 575 95
XM_012961989.2 XP_012817443.1 5 - 6 628 95
GenBank mRNA BC170636.1 - 5 - 6 530 95
BC170638.1 - 5 - 6 530 95
CR762241.2 - 5 - 6 575 95

View Full Product Details