Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
morn5
Entrez Gene ID
548897
Gene Name
MORN repeat containing 5
Gene Aliases
-
Chromosome Location
Chr.8: 16201515 - 16218975 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016143.2 NP_001016143.1 3 - 4 357 107
XM_018096252.1 XP_017951741.1 3 - 4 463 107
GenBank mRNA BC168068.1 - 3 - 4 309 107
BC170640.1 - 3 - 4 337 107
BC170642.1 - 3 - 4 337 107
CR762007.2 - 3 - 4 357 107

View Full Product Details